3-Benzyloxazolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B1339-1G B1339-5G
TCB1339-001GEA 86.8 USD
TCB1339-001G TCB1339-005G
3-Benzyloxazolidine ≥98.0% (by GC, titration analysis)
3-Benzyloxazolidine

CAS Number: 13657-16-4
MDL Number: MFCD00059028
Molecular Formula: C10H13NO
Molecular Weight: 163.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 127
Specific Gravity (20/20): 1.07

Formula: C₁₀H₁₃NO
Boiling Pt: 127 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00059028
CAS Number: 13657-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=603dfede6315acd077d047360bb4c9b7bdc6c2b0&vid=603dfede6315acd077d047360bb4c9b7bdc6c2b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR