4-tert-Butylphthalonitrile ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: 4-(tert-Butyl)phthalonitrile, 4-tert-Butyl-1,2-dicyanobenzene

B1334-5G B1334-25G
TCB1334-005GEA 184.13 USD
TCB1334-005G TCB1334-025G
4-tert-Butylphthalonitrile ≥98.0% (by GC, total nitrogen)
4-tert-Butylphthalonitrile

CAS Number: 32703-80-3
MDL Number: MFCD00060112
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Boiling point (°C): 165
Melting point (°C): 60


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 4-tert-Butyl-1,2-dicyanobenzene

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Boiling Pt: 165 °C
Melting Pt: 60 °C
MDL Number: MFCD00060112
CAS Number: 32703-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80cf7e9b6b6456c632d16632868cd0febfab3061&vid=80cf7e9b6b6456c632d16632868cd0febfab3061&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR