9,10-Bis(chloromethyl)anthracene ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B1327-5G B1327-25G
TCB1327-005GEA 213.62 USD
TCB1327-005G TCB1327-25G
9,10-Bis(chloromethyl)anthracene ≥97.0% (by GC, titration analysis)
9,10-Bis(chloromethyl)anthracene

CAS Number: 10387-13-0
MDL Number: MFCD00045388
Molecular Formula: C16H12Cl2
Molecular Weight: 275.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal

Formula: C₁₆H₁₂Cl₂
MW: 275.17 g/mol
MDL Number: MFCD00045388
CAS Number: 10387-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc31be5badb7fdb44c8138f1af051699c4829e44&vid=fc31be5badb7fdb44c8138f1af051699c4829e44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR