Bis(2,2,2-trifluoroethyl)ether ≥99.0%
Supplier: TCI America
Synonyms:
2,2,2-Trifluoroethyl Ether
B1293-1G
B1293-5G
TCB1293-001GEA
59.91
USD
TCB1293-001G
TCB1293-005G
Bis(2,2,2-trifluoroethyl)ether ≥99.0%
Bis(2,2,2-trifluoroethyl)ether
CAS Number: 333-36-8
MDL Number: MFCD00010604
Molecular Formula: C4H4F6O
Molecular Weight: 182.07
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): 1
Specific Gravity (20/20): 1.42
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Bioscience, Reagents for Central Nervous System Disease Models
Other Product Names / Synonyms: 2,2,2-Trifluoroethyl Ether
Formula:
C₄H₄F₆O MW: 182.07 g/mol Boiling Pt: 64 °C Density: 1.42 (20 °C) Flash Pt: 1 °C |
MDL Number:
MFCD00010604 CAS Number: 333-36-8 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...