tert-Butyl-2-bromopropionate ≥97.0%

Supplier: TCI America

Synonyms: 2-Bromopropionic Acid tert-Butyl Ester, tert-Butyl 2-bromopropanoate

B1277-500G B1277-25G
TCB1277-500GEA 665 USD
TCB1277-500G TCB1277-025G
tert-Butyl-2-bromopropionate ≥97.0%
tert-Butyl-2-bromopropionate

CAS Number: 39149-80-9
MDL Number: MFCD00043897
Molecular Formula: C7H13BrO2
Molecular Weight: 209.08
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Specific Gravity (20/20): 1.26
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Bromopropionic Acid tert-Butyl Ester

Formula: C₇H₁₃BrO₂
Boiling Pt: 62 °C
Density: 1.26 (20 °C)
MDL Number: MFCD00043897
CAS Number: 39149-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e57277f0ef2c3cd5b16b72228fe08d99f0121487&vid=e57277f0ef2c3cd5b16b72228fe08d99f0121487&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR