Lucigenin ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: Bis(N-methylacridinium) nitrate, N,N′-Dimethyl-9,9′-biacridinium dinitrate, 10,10'-Dimethyl-9,9'-biacridinium dinitrate

B1203-1G B1203-5G
TCB1203-001GEA 209.6 USD
TCB1203-001G TCB1203-5G
Lucigenin ≥97.0% (by HPLC)
Lucigenin

[for Chemiluminescence Research]
CAS Number: 2315-97-1
MDL Number: MFCD00011925
Molecular Formula: C28H22N4O6
Molecular Weight: 510.51
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 247


Product Categories: Bioscience, ROS/RNS Detection Reagents, Redox Research Reagents, Analytical Science, Acridinium Salts (Chemiluminescence Reagents)
Other Product Names / Synonyms: Lucigenin, Bis(N-methylacridinium) Nitrate

Formula: C₂₈H₂₂N₄O₆
MW: 510.51 g/mol
Melting Pt: 247 °C
MDL Number: MFCD00011925
CAS Number: 2315-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78e6d6c14fe552ae3e88f95752d0f57715151a78&vid=78e6d6c14fe552ae3e88f95752d0f57715151a78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR