N-tert-Butyldiethanolamin ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2'-(tert-Butylimino)diethanol, N-Bis(2-hydroxyethyl)-tert-butylamine, N-tert-Butyldiethanolamine, 2,2'-(tert-Butylazanediyl)diethanol

B1194-25ML B1194-500ML
TCB1194-025MLEA 37 USD
TCB1194-025ML TCB1194-500ML
N-tert-Butyldiethanolamin ≥97.0% (by titrimetric analysis)
N-tert-Butyldiethanolamin

CAS Number: 2160-93-2
MDL Number: MFCD00020584
Molecular Formula: C8H19NO2
Molecular Weight: 161.25
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 161
Melting point (°C): 49
Flash Point (°C): 110


Other Product Names / Synonyms: N-Bis(2-hydroxyethyl)-tert-butylamine, 2,2'-(tert-Butylimino)diethanol

Formula: C₈H₁₉NO₂
MW: 161.2420 g/mol
Boiling Pt: 161 °C
Melting Pt: 49 °C
Flash Pt: 110 °C
MDL Number: MFCD00020584
CAS Number: 2160-93-2
EINECS: 218-480-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fe53fb337352e422b3d10ed4f952f66b0aef453&vid=6fe53fb337352e422b3d10ed4f952f66b0aef453&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR