4-Bromopyridine hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1171-5G B1171-25G
TCB1171-005GEA 63 USD
TCB1171-005G TCB1171-025G
4-Bromopyridine hydrobromide ≥98.0% (by titrimetric analysis)
4-Bromopyridine hydrobromide

CAS Number: 74129-11-6
MDL Number: MFCD00060195
Molecular Formula: C5H4BrN
Molecular Weight: 238.91
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C

Formula: C₅H₄BrN
MDL Number: MFCD00060195
CAS Number: 74129-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaf3ed3b44d252d1adc01dd80d01f836c956ee14&vid=eaf3ed3b44d252d1adc01dd80d01f836c956ee14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR