1-Bromo-2-methoxymethylnaphthalene ≥98.0%

Supplier: TCI America

Synonyms: 1-Bromo-2-(methoxymethyl)naphthalene

B1170-5G B1170-25G
TCB1170-005GEA 159 USD
TCB1170-005G TCB1170-025G
1-Bromo-2-methoxymethylnaphthalene ≥98.0%
1-Bromo-2-methoxymethylnaphthalene

CAS Number: 64689-70-9
MDL Number: MFCD00059518
Molecular Formula: C12H11BrO
Molecular Weight: 251.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Specific Gravity (20/20): 1.43

Formula: C₁₂H₁₁BrO
MW: 251.12 g/mol
Boiling Pt: 151 °C
Density: 1.43 (20 °C)
MDL Number: MFCD00059518
CAS Number: 64689-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d19a514675d003858d1f22d227965a49594d5b7&vid=7d19a514675d003858d1f22d227965a49594d5b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR