2-Amino-4-nitrophenol sodium salt ≥80.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Amino-4-nitrophenol Sodium Salt, Sodium 2-amino-4-nitrophenolate, 2-Hydroxy-5-nitroaniline Sodium Salt

A0380-500G A0380-25G
TCA0380-500GEA 139 USD
TCA0380-500G TCA0380-025G
2-Amino-4-nitrophenol sodium salt ≥80.0% (by titrimetric analysis)
2-Amino-4-nitrophenol sodium salt

CAS Number: 61702-43-0
MDL Number: MFCD00067428
Molecular Formula: C6H6N2O3
Molecular Weight: 176.11
Purity/Analysis Method: >80.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Hydroxy-5-nitroaniline Sodium Salt

Formula: C₆H₆N₂O₃
MW: 176.11 g/mol
MDL Number: MFCD00067428
CAS Number: 61702-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0831c24f55c4191234db5d14965f151955cda459&vid=0831c24f55c4191234db5d14965f151955cda459&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >80.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR