N-(Butoxymethyl)acrylamide ≥96.0% (by titrimetric analysis) stabilized
Supplier: TCI America
B1060-25ML
B1060-500ML
TCB1060-025MLEA
28
USD
TCB1060-025ML
TCB1060-500ML
N-(Butoxymethyl)acrylamide ≥96.0% (by titrimetric analysis) stabilized
N-(Butoxymethyl)acrylamide
(stabilized with MEHQ)
CAS Number: 1852-16-0
MDL Number: MFCD00059417
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method:
Formula:
C₈H₁₅NO₂ Density: 0.98 (20 °C) Flash Pt: 78 °C |
MDL Number:
MFCD00059417 CAS Number: 1852-16-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...