1-Bromopropene ≥97.0%

Supplier: TCI America

Synonyms: 1-Bromo-1-propene, Propenyl bromide

B1038-25G B1038-5G
TCB1038-025GEA 238 USD
TCB1038-025G TCB1038-5G
1-Bromopropene ≥97.0%
1-Bromopropene

CAS Number: 590-14-7
MDL Number: MFCD00000186
Molecular Formula: C3H5Br
Molecular Weight: 120.98
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Melting point (°C): -116
Flash Point (°C): 4
Specific Gravity (20/20): 1.43
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Propenyl Bromide

Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 63 °C
Melting Pt: –116 °C
Density: 1.43 (20 °C)
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000186
CAS Number: 590-14-7
EINECS: 209-671-6
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cc412970613a3897dff8737be2769ffe900b632&vid=f137181da5ff35a411ff8ca1b185effb2e49a363&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR