6-tert-Butyl-2,4-xylenol ≥97.0%
Supplier: TCI America
Synonyms:
2-(tert-Butyl)-4,6-dimethylphenol
B0903-25G
B0903-500G
B0903-100G
TCB0903-025GEA
47.01
USD
TCB0903-025G
TCB0903-500G
TCB0903-100G
6-tert-Butyl-2,4-xylenol ≥97.0%
6-tert-Butyl-2,4-xylenol
CAS Number: 1879-09-0
MDL Number: MFCD00002234
Molecular Formula: C12H18O
Molecular Weight: 178.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 249
Flash Point (°C): 112
Freezing point (°C): 22
Other Product Names / Synonyms: 2-tert-Butyl-4,6-dimethylphenol
Formula:
C₁₂H₁₈O MW: 178.28 g/mol Boiling Pt: 249 °C Flash Pt: 112 °C |
MDL Number:
MFCD00002234 CAS Number: 1879-09-0 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...