Boron trifluoridedimethanol complex
Supplier: TCI America
B0894-100ML
B0894-500ML
TCB0894-100MLEA
30.3
USD
TCB0894-100ML
TCB0894-500ML
Boron trifluoridedimethanol complex
Boron trifluoridedimethanol complex
CAS Number: 2802-68-8
Molecular Formula: C2H8BF3O2
Molecular Weight: 131.89
Form: Liquid
Color: Colorless
Flash Point (°C): 29
Specific Gravity (20/20): 1.22
Formula:
BF₃·2CH₃OH MW: 131.89 g/mol |
MDL Number:
MFCD00071635 CAS Number: 2802-68-8 |
|
Specification Test Results
Concentration (Gravimetric Method) | 49.0 to 54.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...