(±)-1-Butoxy-2-propanol ≥96.0%

Supplier: TCI America
Warning

Synonyms: (±)-Monopropylene glycol butyl ether, Propylene glycol butyl ether, Solvent 35, 1-Butoxy-2-propanol, 1-Butoxypropan-2-ol, Propylene glycol 1-Monobutyl ether, (±)-Propylene glycol monobutyl ether

B0864-25ML B0864-500ML
TCB0864-025MLEA 33.75 USD
TCB0864-025ML TCB0864-500ML
(±)-1-Butoxy-2-propanol ≥96.0%
(±)-1-Butoxy-2-propanol

CAS Number: 5131-66-8
MDL Number: MFCD00021904
Molecular Formula: C7H16O2
Molecular Weight: 132.20
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 62
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Propylene Glycol 1-Monobutyl Ether

Formula: C₇H₁₆O₂
MW: 132.2 g/mol
Boiling Pt: 170 °C
Density: 0.88 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00021904
CAS Number: 5131-66-8
EINECS: 225-878-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c872dc1269c2204cd3d79c7b09c6dbd6bc991a30&vid=c872dc1269c2204cd3d79c7b09c6dbd6bc991a30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR