Bis(chloromethyl)sulfide ≥97.0%
Supplier: TCI America
Synonyms:
Dichlorodimethyl sulphide, Di(chloromethyl) sulphide, Chloromethyl Sulphide
B0843-5G
B0843-25G
TCB0843-005GEA
209.61
USD
TCB0843-005G
TCB0843-025G
Bis(chloromethyl)sulfide ≥97.0%
Bis(chloromethyl)sulfide
CAS Number: 3592-44-7
MDL Number: MFCD00059474
Molecular Formula: C2H4Cl2S
Molecular Weight: 131.01
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Specific Gravity (20/20): 1.41
Other Product Names / Synonyms: Chloromethyl Sulfide, Dichlorodimethyl Sulfide, Di(chloromethyl) Sulfide
Formula:
C₂H₄Cl₂S MW: 131.01 g/mol Boiling Pt: 156 °C Density: 1.41 (20 °C) |
MDL Number:
MFCD00059474 CAS Number: 3592-44-7 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...