Di-tert-butyl sulfide ≥98.0%

Supplier: TCI America

B0807-25ML B0807-500ML
TCB0807-025MLEA 90.2 USD
TCB0807-025ML TCB0807-500ML
Di-tert-butyl sulfide ≥98.0%
Di-tert-butyl sulphide

CAS Number: 107-47-1
MDL Number: MFCD00008839
Molecular Formula: C8H18S
Molecular Weight: 146.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 26
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Di-tert-butyl Sulfide

Formula: C₈H₁₈S
Boiling Pt: 150 °C
Density: 0.83 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00008839
CAS Number: 107-47-1
EINECS: 203-493-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0af8e0e23ba379323e8a29978de0a3b158b9eab1&vid=0af8e0e23ba379323e8a29978de0a3b158b9eab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR