Vinyl butyrate ≥98.0% stabilized

Supplier: TCI America

B0765-25ML B0765-500ML B0765-100ML
TCB0765-025MLEA 19 USD
TCB0765-025ML TCB0765-500ML TCB0765-100ML
Vinyl butyrate ≥98.0% stabilized
Vinyl butyrate

(stabilized with MEHQ)
CAS Number: 123-20-6
MDL Number: MFCD00059410
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 20
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Butyric Acid Vinyl Ester

Formula: C₆H₁₀O₂
Boiling Pt: 116 °C
Density: 0.90 (20 °C)
Flash Pt: 20 °C
MDL Number: MFCD00059410
CAS Number: 123-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa06d437ff5cb6a65ef71849c7515a244e120905&vid=fa06d437ff5cb6a65ef71849c7515a244e120905&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR