N-Butyl-p-toluenesulfonamide ≥97.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
N-Butyl-4-methylbenzenesulphonamide
B0745-500G
B0745-25G
TCB0745-500GEA
526
USD
TCB0745-500G
TCB0745-025G
N-Butyl-p-toluenesulfonamide ≥97.0% (by total nitrogen basis)
N-Butyl-p-toluenesulfonamide
CAS Number: 1907-65-9
MDL Number: MFCD00027162
Molecular Formula: C11H17NO2S
Molecular Weight: 227.32
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Boiling point (°C): 234
Melting point (°C): 41
Formula:
C₁₁H₁₇NO₂S MW: 227.32 g/mol Boiling Pt: 234 °C Melting Pt: 41 °C |
MDL Number:
MFCD00027162 CAS Number: 1907-65-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...