Tetrabutyl orthotitanate tetramer
Supplier: TCI America
Synonyms:
Butyl Titanate Tetramer, Tetrabutoxy Titan Tetramer, Tetrabutyl Titanate Tetramer, Titanium Butoxide Tetramer
B0743-500G
B0743-25G
B0743-100G
TCB0743-500GEA
267
USD
TCB0743-500G
TCB0743-025G
TCB0743-100G
Tetrabutyl orthotitanate tetramer
Tetrabutyl orthotitanate tetramer
CAS Number: 70799-68-7
MDL Number: MFCD00070567
Molecular Formula: C16H36O4Ti
Molecular Weight: 970.62
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 50
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: Butyl Titanate Tetramer, Tetrabutoxy Titan Tetramer, Tetrabutyl Titanate Tetramer, Titanium Butoxide Tetramer
Formula:
C₁₆H₃₆O₄Ti MW: 970.62 g/mol Density: 1.10 (20 °C) Flash Pt: 50 °C |
MDL Number:
MFCD00070567 CAS Number: 70799-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...