Titanium(IV) butanolate

Supplier: TCI America
Danger

B0742-25G B0742-500G
TCB0742-025GEA 20.34 USD
TCB0742-025G TCB0742-500G
Titanium(IV) butanolate
Titanium(IV) butanolate

CAS Number: 5593-70-4
MDL Number: MFCD00009433
Molecular Formula: C16H36O4Ti
Molecular Weight: 340.33
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 185
Melting point (°C): -40
Flash Point (°C): 46
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Butyl Titanate, Tetrabutoxy Titan, Tetrabutyl Titanate, Titanium Butoxide

Formula: C₁₆H₃₆O₄Ti
Boiling Pt: 185 °C
Melting Pt: –40 °C
Density: 1.00 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00009433
CAS Number: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51643979ff74e929fd24a23769e2908f61470110&vid=51643979ff74e929fd24a23769e2908f61470110&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR