2-tert-Butyl-4-methoxyphenol ≥98.0%

Supplier: TCI America

Synonyms: 3-BHA, 4-Hydroxy-3-tert-butylanisole, 3-tert-Butyl-4-hydroxyanisole

B0723-25G B0723-500G B0723-100G
TCB0723-025GEA 31.54 USD
TCB0723-025G TCB0723-500G TCB0723-100G
2-tert-Butyl-4-methoxyphenol ≥98.0%
2-tert-Butyl-4-methoxyphenol

CAS Number: 121-00-6
MDL Number: MFCD00040484
Molecular Formula: C11H16O2
Molecular Weight: 180.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 60
Flash Point (°C): 130


Other Product Names / Synonyms: 2-tert-Butyl-4-methoxyphenol

Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Boiling Pt: 267…269 °C (1013 hPa)
Melting Pt: 60 °C
Density: 1.0587 g/cm³ (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD00040484
CAS Number: 121-00-6
EINECS: 204-442-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bfe7fb9b5df5474922a8254db593e89b793e93d&vid=4bfe7fb9b5df5474922a8254db593e89b793e93d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR