1,2-Epoxybutane ≥99.0%

Supplier: TCI America

Synonyms: 1,2-Butylene oxide, 1,2-Butylene Oxide, 1,2-Epoxybutane, Ethylethylene Oxide, Ethyloxirane

B0719-25ML B0719-500ML B0719-100ML
TCB0719-025MLEA 19.05 USD
TCB0719-025ML TCB0719-500ML TCB0719-100ML
1,2-Epoxybutane ≥99.0%
1,2-Epoxybutane

CAS Number: 106-88-7
MDL Number: MFCD00005153
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Flash Point (°C): -22
Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Epoxybutane, Ethylethylene Oxide, Ethyloxirane

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 63 °C
Melting Pt: –130 °C
Density: 0.83 (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00005153
CAS Number: 106-88-7
EINECS: 203-438-2
UN: 3022
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff51ece749bb292a8923432b669479843963af95&vid=ff51ece749bb292a8923432b669479843963af95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR