N-Butylbenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-Benzenesulphonylbutylamine

B0716-25G B0716-500G
TCB0716-025GEA 20.02 USD
TCB0716-025G TCB0716-500G
N-Butylbenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
N-Butylbenzenesulfonamide

CAS Number: 3622-84-2
MDL Number: MFCD00025024
Molecular Formula: C10H15NO2S
Molecular Weight: 213.30
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Clear Liquid
Boiling point (°C): 184
Flash Point (°C): 118
Specific Gravity (20/20): 1.15


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: N-Benzenesulfonylbutylamine

Formula: C₁₀H₁₅NO₂S
MW: 213.30 g/mol
Boiling Pt: 184 °C
Density: 1.15 (20 °C)
Flash Pt: 118 °C
MDL Number: MFCD00025024
CAS Number: 3622-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd1534ca610caa087b3e804e89ae8638e45955d4&vid=fd1534ca610caa087b3e804e89ae8638e45955d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR