Butyl glycidyl ether ≥98.0%
Supplier: TCI America
B0697-25ML
B0697-500ML
TCB0697-025MLEA
45.76
USD
TCB0697-025ML
TCB0697-500ML
Butyl glycidyl ether ≥98.0%
Butyl glycidyl ether
CAS Number: 2426-08-6
MDL Number: MFCD00005145
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 59
Specific Gravity (20/20): 0.92
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1-Butoxy-2,3-epoxypropane
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 167 °C Density: 0.92 (20 °C) Flash Pt: 59 °C |
MDL Number:
MFCD00005145 CAS Number: 2426-08-6 EINECS: 219-376-4 UN: 3271 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...