Undecanoyl chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Undecanoyl Chloride, Undecanoylchloride

U0026-10ML U0026-25ML
TCU0026-010MLEA 81 USD
TCU0026-010ML TCU0026-025ML
Undecanoyl chloride ≥97.0% (by titrimetric analysis)
Undecanoyl chloride

CAS Number: 17746-05-3
MDL Number: MFCD00000739
Molecular Formula: C11H21ClO
Molecular Weight: 204.74
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 130
Flash Point (°C): 110
Specific Gravity (20/20): 0.93

Formula: C₁₁H₂₁ClO
MW: 204.74 g/mol
Boiling Pt: 130 °C
Density: 0.93 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000739
CAS Number: 17746-05-3
EINECS: 241-741-1
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1bf05feba7a47186c74ae90094170e6196777d4&vid=b1bf05feba7a47186c74ae90094170e6196777d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR