1,1,2,2-Tetraphenyldisilane ≥97.0%
Supplier: TCI America
CAS Number: 16343-18-3
MDL Number: MFCD00093170
Molecular Formula: C24H22Si2
Molecular Weight: 366.61
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 80
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: TPDS
Formula:
C₂₄H₂₂Si₂ Melting Pt: 80 °C |
MDL Number:
MFCD00093170 CAS Number: 16343-18-3 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...