1,1,2,2-Tetrachloro-1,2-dimethyldisilane ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1590-5G T1590-25G
TCT1590-005GEA 206 USD
TCT1590-005G TCT1590-25G
1,1,2,2-Tetrachloro-1,2-dimethyldisilane ≥98.0% (by titrimetric analysis)
1,1,2,2-Tetrachloro-1,2-dimethyldisilane

CAS Number: 4518-98-3
MDL Number: MFCD00192469
Molecular Formula: C2H6Cl4Si2
Molecular Weight: 228.04
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 49
Storage Temperature: 0-10°C

Formula: C₂H₆Cl₄Si₂
Flash Pt: 49 °C
MDL Number: MFCD00192469
CAS Number: 4518-98-3
EINECS: 224-844-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba7b953a1101de2e887729e216cd053d5bc570ac&vid=ba7b953a1101de2e887729e216cd053d5bc570ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR