Triphenylacetic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,2,2-Triphenylacetic acid
T1585-25G
T1585-5G
TCT1585-25GEA
521
USD
TCT1585-25G
TCT1585-005G
Triphenylacetic acid ≥97.0% (by titrimetric analysis)
Triphenylacetic acid
CAS Number: 595-91-5
MDL Number: MFCD00004185
Molecular Formula: C20H16O2
Molecular Weight: 288.35
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 270
Formula:
C₂₀H₁₆O₂ MW: 288.35 g/mol Boiling Pt: 437 °C (1013 hPa) Melting Pt: 270 °C Density: 1.27 g/cm³ (20 °C) |
MDL Number:
MFCD00004185 CAS Number: 595-91-5 EINECS: 209-873-4 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...