2,3,5-Trichloropyridine ≥98.0%

Supplier: TCI America

Synonyms: 2,3,5-TCP

T1479-25G T1479-250G
TCT1479-025GEA 36 USD
TCT1479-025G TCT1479-250G
2,3,5-Trichloropyridine ≥98.0%
2,3,5-Trichloropyridine

CAS Number: 16063-70-0
MDL Number: MFCD00043007
Molecular Formula: C5H2Cl3N
Molecular Weight: 182.43
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 220
Melting point (°C): 47
Flash Point (°C): 110

Formula: C₅H₂Cl₃N
MW: 182.44 g/mol
Boiling Pt: 220 °C
Melting Pt: 47 °C
Flash Pt: 110 °C
MDL Number: MFCD00043007
CAS Number: 16063-70-0
EINECS: 407-270-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcb7e9cdb83095f7782c36042061d101d8717298&vid=dcb7e9cdb83095f7782c36042061d101d8717298&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR