2,2,6,6-Tetramethyl-4-piperidone ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Triacetonamine, 2,2,6,6-Tetramethylpiperidin-4-one
T1424-25G
T1424-500G
T1424-100G
TCT1424-025GEA
35.73
USD
TCT1424-025G
TCT1424-500G
TCT1424-100G
2,2,6,6-Tetramethyl-4-piperidone ≥98.0% (by titrimetric analysis)
2,2,6,6-Tetramethyl-4-piperidone
CAS Number: 826-36-8
MDL Number: MFCD00149393
Molecular Formula: C9H17NO
Molecular Weight: 155.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 105
Melting point (°C): 38
Flash Point (°C): 73
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Triacetonamine
Formula:
C₉H₁₇NO MW: 155.24 g/mol Boiling Pt: 105 °C Melting Pt: 38 °C Flash Pt: 73 °C |
MDL Number:
MFCD00149393 CAS Number: 826-36-8 EINECS: 212-554-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...