Tetrafluorophthalonitrile ≥98.0%
Supplier: TCI America
Synonyms:
3,4,5,6-Tetrafluorophthalonitrile, 1,2-Dicyano-3,4,5,6-tetrafluorobenzene
T1402-25G
T1402-5G
TCT1402-25GEA
141
USD
TCT1402-25G
TCT1402-005G
Tetrafluorophthalonitrile ≥98.0%
Tetrafluorophthalonitrile
CAS Number: 1835-65-0
MDL Number: MFCD00001774
Molecular Formula: C8F4N2
Molecular Weight: 200.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 87
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,2-Dicyano-3,4,5,6-tetrafluorobenzene
Formula:
C₈F₄N₂ MW: 200.1 g/mol Melting Pt: 87 °C |
MDL Number:
MFCD00001774 CAS Number: 1835-65-0 EINECS: 217-400-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...