2,3,5-Trimethylthiophene ≥98.0%

Supplier: TCI America

T1327-1G T1327-5G
TCT1327-001GEA 116 USD
TCT1327-001G TCT1327-005G
2,3,5-Trimethylthiophene ≥98.0%
2,3,5-Trimethylthiophene

CAS Number: 1795-05-7
MDL Number: MFCD00142784
Molecular Formula: C7H10S
Molecular Weight: 126.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 82
Specific Gravity (20/20): 0.98

Formula: C₇H₁₀S
Boiling Pt: 82 °C
Density: 0.98 (20 °C)
MDL Number: MFCD00142784
CAS Number: 1795-05-7
EINECS: 217-269-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acc995956ad3ed1e262f313881038256e542aaa4&vid=acc995956ad3ed1e262f313881038256e542aaa4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR