Tris(diethylamino)phosphine ≥90.0%

Supplier: TCI America

Synonyms: Hexaethyltriaminophosphine, Hexaethylphosphorus triamide, Hexaethyl phosphorous triamide

T1309-25ML T1309-5ML
TCT1309-025MLEA 187 USD
TCT1309-025ML TCT1309-005ML
Tris(diethylamino)phosphine ≥90.0%
Tris(diethylamino)phosphine

CAS Number: 2283-11-6
MDL Number: MFCD00009041
Molecular Formula: C12H30N3P
Molecular Weight: 247.37
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 245
Flash Point (°C): 26
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Hexaethylphosphorous Triamide

Formula: C₁₂H₃₀N₃P
MW: 247.36 g/mol
Boiling Pt: 245 °C
Density: 0.90 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00009041
CAS Number: 2283-11-6
EINECS: 218-920-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97e24f43b66a4f76624136459122f3abfbc66d23&vid=97e24f43b66a4f76624136459122f3abfbc66d23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR