Tetrakis(dimethylamino)ethylene ≥95.0%

Supplier: TCI America

Synonyms: TDAE

T1221-25G T1221-5G
TCT1221-25GEA 4830 USD
TCT1221-25G TCT1221-5G
Tetrakis(dimethylamino)ethylene ≥95.0%
Tetrakis(dimethylamino)ethylene

CAS Number: 996-70-3
MDL Number: MFCD00009601
Molecular Formula: C10H24N4
Molecular Weight: 200.33
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 59
Flash Point (°C): 53
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: TDAE

Formula: C₁₀H₂₄N₄
MW: 200.33 g/mol
Boiling Pt: 59 °C
Density: 0.87 (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00009601
CAS Number: 996-70-3
EINECS: 213-638-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8fe4871059c33251b828c4fd9fdb3fe2003a164&vid=a8fe4871059c33251b828c4fd9fdb3fe2003a164&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR