2,3,6-Trichlorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T1114-1G T1114-10G
TCT1114-001GEA 105 USD
TCT1114-001G TCT1114-010G
2,3,6-Trichlorophenol ≥98.0% (by GC, titration analysis)
2,3,6-Trichlorophenol

CAS Number: 933-75-5
MDL Number: MFCD00002165
Molecular Formula: C6H3Cl3O
Molecular Weight: 197.44
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 250
Melting point (°C): 55
Flash Point (°C): 78

Formula: C₆H₃Cl₃O
Boiling Pt: 250 °C
Melting Pt: 55 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00002165
CAS Number: 933-75-5
EINECS: 213-271-7
UN: 2020
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7a748e5b4d8379b3e137bc0d82f11c952e1a506&vid=b77ec4b4d7bfd14ec9d6cadbffc4c3576627aacb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR