Tetrafluoroterephthalonitrile ≥98.0%
Supplier: TCI America
Synonyms:
Perfluoroterephthalonitrile
T1050-5G
T1050-1G
T1050-25G
TCT1050-005GEA
28
USD
TCT1050-005G
TCT1050-001G
TCT1050-25G
Tetrafluoroterephthalonitrile ≥98.0%
Tetrafluoroterephthalonitrile
CAS Number: 1835-49-0
MDL Number: MFCD00001776
Molecular Formula: C8F4N2
Molecular Weight: 200.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 198
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluoroterephthalonitrile
Formula:
C₈F₄N₂ MW: 200.1 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00001776 CAS Number: 1835-49-0 EINECS: 217-397-3 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...