1,2,4,5-Tetracyanobenzene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Pyromelliticnitrile, 1,2,4,5-Benzenetetranitrile, Benzene-1,2,4,5-tetracarbonitrile, 1,2,4,5-Benzenetetracarbonitrile, Benzene-1,2,4,5-tetracarbonitrile, Pyromellitotetranitrile

T0988-1G T0988-5G
TCT0988-001GEA 78.5 USD
TCT0988-001G TCT0988-005G
1,2,4,5-Tetracyanobenzene ≥98.0% (by HPLC, total nitrogen)
1,2,4,5-Tetracyanobenzene

CAS Number: 712-74-3
MDL Number: MFCD00043481
Molecular Formula: C10H2N4
Molecular Weight: 178.15
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 265

Formula: C₁₀H₂N₄
MW: 178.15 g/mol
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 265 °C
Density: 1.372 g/cm³ (20 °C)
MDL Number: MFCD00043481
CAS Number: 712-74-3
UN: 3439
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec91df520bad43e553c6a517bca33adf64c8ce93&vid=ec91df520bad43e553c6a517bca33adf64c8ce93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR