3,5,5-Trimethylhexanal ≥90.0%

Supplier: TCI America
Danger

T0940-500ML T0940-25ML
TCT0940-500MLEA 99.4 USD
TCT0940-500ML TCT0940-025ML
3,5,5-Trimethylhexanal ≥90.0%
3,5,5-Trimethylhexanal

CAS Number: 5435-64-3
MDL Number: MFCD00010441
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 167
Flash Point (°C): 46
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 3,5,5-Trimethylhexanaldehyde

Formula: C₉H₁₈O
Boiling Pt: 167 °C
Density: 0.82 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00010441
CAS Number: 5435-64-3
EINECS: 226-603-0
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=326f2931cc7abc228c67e22d7f07a7492af86399&vid=326f2931cc7abc228c67e22d7f07a7492af86399&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR