Tetrabutylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: Tetrabutylammoniumtetrafluoroborate

T0914-500G T0914-25G T0914-100G
TCT0914-500GEA 299.13 USD
TCT0914-500G TCT0914-025G TCT0914-100G
Tetrabutylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Tetrabutylammonium tetrafluoroborate

CAS Number: 429-42-5
MDL Number: MFCD00011634
Molecular Formula: C16H36BF4N
Molecular Weight: 329.27
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 161

Formula: C₁₆H₃₆BF₄N
MW: 329.27 g/mol
Melting Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS Number: 429-42-5
EINECS: 207-058-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=666d9446c436bab91e308061de79c9357ebb3a76&vid=666d9446c436bab91e308061de79c9357ebb3a76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR