2,3,6-Trimethylphenol ≥98.0%
Supplier: TCI America
Synonyms:
3-Hydroxypseudocumene
T0877-25G
T0877-500G
TCT0877-025GEA
23
USD
TCT0877-025G
TCT0877-500G
2,3,6-Trimethylphenol ≥98.0%
2,3,6-Trimethylphenol
CAS Number: 2416-94-6
MDL Number: MFCD00002229
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 120
Melting point (°C): 62
Flash Point (°C): 108
Other Product Names / Synonyms: 3-Hydroxypseudocumene
Formula:
C₉H₁₂O MW: 136.19 g/mol Boiling Pt: 120 °C Melting Pt: 62 °C Density: 1.189 g/cm³ (20 °C) Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002229 CAS Number: 2416-94-6 EINECS: 219-330-3 UN: 2430 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...