1,2,3,5-Tetrachlorobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
T0870-25G
T0870-5G
TCT0870-25GEA
426
USD
TCT0870-25G
TCT0870-5G
1,2,3,5-Tetrachlorobenzene ≥98.0%
1,2,3,5-Tetrachlorobenzene
CAS Number: 634-90-2
MDL Number: MFCD00000543
Molecular Formula: C6H2Cl4
Molecular Weight: 215.88
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 246
Melting point (°C): 52
Flash Point (°C): 155
Formula:
C₆H₂Cl₄ MW: 215.89 g/mol Boiling Pt: 246 °C Melting Pt: 52 °C Density: 1.858 g/cm³ (25 °C) Flash Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000543 CAS Number: 634-90-2 EINECS: 211-217-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...