Tetraethylammonium fluoride hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetrylammonium fluoride hydrate, Tetraethylammonium fluoride xhydrate

T0838-25G T0838-5G
TCT0838-025GEA 395.83 USD
TCT0838-025G TCT0838-005G
Tetraethylammonium fluoride hydrate ≥98.0% (by titrimetric analysis)
Tetraethylammonium fluoride hydrate

CAS Number: 665-46-3
MDL Number: MFCD00011826
Molecular Formula: C8H20FN
Molecular Weight: 149.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₈H₂₀FN·xH₂O
MW: 149.25 g/mol (anhy)
MDL Number: MFCD00149990
CAS Number: 98330-04-2
EINECS: 211-559-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82076652f3608416a0007ecd9f2689dc8171dfb5&vid=82076652f3608416a0007ecd9f2689dc8171dfb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR