1,1,2-Trimethoxyethane ≥98.0%

Supplier: TCI America

Synonyms: Methoxyacetaldehyde dimethyl Acetal

T0831-25ML T0831-100ML
TCT0831-025MLEA 168 USD
TCT0831-025ML TCT0831-100ML
1,1,2-Trimethoxyethane ≥98.0%
1,1,2-Trimethoxyethane

CAS Number: 24332-20-5
MDL Number: MFCD00008486
Molecular Formula: C5H12O3
Molecular Weight: 120.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 126
Flash Point (°C): 23
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Methoxyacetaldehyde Dimethyl Acetal

Formula: C₅H₁₂O₃
MW: 120.1470 g/mol
Boiling Pt: 126 °C
Density: 0.96 (20 °C)
Flash Pt: 23 °C
MDL Number: MFCD00008486
CAS Number: 24332-20-5
EINECS: 246-175-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aaa3dcfaba7c77ccbfb1f38b62ba6c4a3c5e5a6f&vid=aaa3dcfaba7c77ccbfb1f38b62ba6c4a3c5e5a6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR