DL-m-Tyrosine ≥98.0%
Supplier: TCI America
Synonyms:
3-Hydroxyphenylalanine, 3-(m-Hidroxifenil)-DL-alanina, 3-(3-Hydroxyphenyl)-DL-alanine
T0787-5G
T0787-1G
TCT0787-5GEA
182
USD
TCT0787-5G
TCT0787-001G
DL-m-Tyrosine ≥98.0%
DL-m-Tyrosine
CAS Number: 775-06-4
MDL Number: MFCD00002597
Molecular Formula: C9H11NO3
Molecular Weight: 181.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 285
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 3-(3-Hydroxyphenyl)-DL-alanine
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol Boiling Pt: 351 Melting Pt: 283 Density: 1.456 g/cm³ (20 °C) |
MDL Number:
MFCD00002597 CAS Number: 775-06-4 EINECS: 212-270-9 Merck Index: 12,09971 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...