2,2,3-Trimethylbutane ≥96.0%
Supplier: TCI America
|
Danger
|
T0711-5ML
T0711-25ML
TCT0711-005MLEA
86
USD
TCT0711-005ML
TCT0711-025ML
2,2,3-Trimethylbutane ≥96.0%
2,2,3-Trimethylbutane
CAS Number: 464-06-2
MDL Number: MFCD00039846
Molecular Formula: C7H16
Molecular Weight: 100.21
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 81
Melting point (°C): -24
Flash Point (°C): -20
Specific Gravity (20/20): 0.69
Formula:
C₇H₁₆ MW: 100.2 g/mol Boiling Pt: 81 °C Melting Pt: -24 °C Density: 0.69 (20 °C) Flash Pt: -20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039846 CAS Number: 464-06-2 EINECS: 207-346-3 UN: 1206 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...