Phthalylsulfathiazole ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Phthalylsulphathiazole, 2-((4-(N-(Thiazol-2-yl)sulphamoyl)phenyl)carbamoyl)benzoic acid, 4'-(2-Thiazolylsulphamoyl)phthalanilic acid
T0703-500G
T0703-25G
TCT0703-500GEA
193.5
USD
TCT0703-500G
TCT0703-025G
Phthalylsulfathiazole ≥95.0% (by titrimetric analysis)
Phthalylsulphathiazole
CAS Number: 85-73-4
MDL Number: MFCD00005318
Molecular Formula: C17H13N3O5S2
Molecular Weight: 403.43
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 240
Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 4'-(2-Thiazolylsulfamoyl)phthalanilic Acid
Formula:
C₁₇H₁₃N₃O₅S₂ MW: 403.44 g/mol Melting Pt: 240 °C |
MDL Number:
MFCD00005318 CAS Number: 85-73-4 EINECS: 201-627-4 |
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...