2,3,3-Trimethyl-1-butene ≥98.0%

Supplier: TCI America

T0671-5ML T0671-1ML
TCT0671-005MLEA 164 USD
TCT0671-005ML TCT0671-1ML
2,3,3-Trimethyl-1-butene ≥98.0%
2,3,3-Trimethyl-1-butene

CAS Number: 594-56-9
MDL Number: MFCD00008850
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 78
Flash Point (°C): -17
Specific Gravity (20/20): 0.71

Formula: C₇H₁₄
MW: 98.19 g/mol
Boiling Pt: 78 °C
Melting Pt: -109.9 °C
Density: 0.71 (20 °C)
Flash Pt: -17 °C
MDL Number: MFCD00008850
CAS Number: 594-56-9
EINECS: 209-845-1
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a4820d4b82767c4b7b61c6c99034cc9a383f1b7&vid=2d0ae83c690f6c0b369be283e30f090cc5758057&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR