2,4,6-Tris(dimethylaminomethyl)phenol ≥80.0%

Supplier: TCI America

T0526-500G T0526-25G
TCT0526-500GEA 112 USD
TCT0526-500G TCT0526-025G
2,4,6-Tris(dimethylaminomethyl)phenol ≥80.0%
2,4,6-Tris(dimethylaminomethyl)phenol

CAS Number: 90-72-2
MDL Number: MFCD00008330
Molecular Formula: C15H27N3O
Molecular Weight: 265.40
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 135
Flash Point (°C): 124
Specific Gravity (20/20): 0.98

Formula: C₁₅H₂₇N₃O
Boiling Pt: 135 °C
Density: 0.98 (20 °C)
Flash Pt: 124 °C
MDL Number: MFCD00008330
CAS Number: 90-72-2
EINECS: 202-013-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cae4a8195c3e104bba3e90b46db0aa72fb8c25&vid=13cae4a8195c3e104bba3e90b46db0aa72fb8c25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR