1,2,3-Trimethylbenzene ≥80.0%
Supplier: TCI America
CAS Number: 526-73-8
MDL Number: MFCD00008520
Molecular Formula: C9H12
Molecular Weight: 120.20
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 176
Flash Point (°C): 44
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: Hemimellitene
Formula:
C₉H₁₂ Boiling Pt: 176 °C Melting Pt: –25 °C Density: 0.89 (20 °C) Flash Pt: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008520 CAS Number: 526-73-8 EINECS: 208-394-8 UN: 3295 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >80.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...